Order Entry
Italy
ContactUsLinkComponent
Zinc, granular 20 mesh
Flammable
Environment
Zinc, granular 20 mesh
Certificates
undefined
Flammable
Environment
Zinc, granular 20 mesh
Certificates
Specification Test Results
  • Appearance:
    Silver grey Granules or filings
  • Titration Complexometric:
    ≥99.8 %
  • Particle size:
    20 mesh
  • Suitability:
    (For determination of As) Passes test
  • Iron (Fe):
    ≤0.01 %
  • Lead (Pb):
    ≤0.01 %