Order Entry
Italy
ContactUsLinkComponent
 
Iodine 99.5-100.5%, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent, resublimed
 
undefined
Iodine 99.5-100.5%, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent, resublimed
 

 

  • Assay:
    99.5 - 100.5 %
  • Identification A:
    Passes test Ph.Eur.
  • Identification B:
    Passes test Ph.Eur.
  • Insoluble substances:
    Passes test
  • Non-volatile residue:
    Max. 0.02 %
  • Cl + Br (as Cl):
    Max. 30 ppm
  • SO4 (Sulphate):
    Max. 50 ppm
  • Conforms to Reag. Ph.Eur.:
    Passes test

 

Frequently Bought Together