Order Entry
Italy
ContactUsLinkComponent
Bromine for synthesis, Sigma-Aldrich®
Toxic
Corrosive
Environment
Bromine for synthesis, Sigma-Aldrich®
Catalog # 8.20171.1000
Supplier:  Merck
CAS Number:  
Toxic
Corrosive
Environment
Bromine for synthesis, Sigma-Aldrich®
Catalog # 8.20171.1000
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle, plastic-coated, returnable
  • Pk:
    1 L
  • Seq:
    5037

Specifications

About this item

 :
 :159.81 g/mol
Merck Index:13,01378
Boiling Pt:58.7 °C (1013 hPa)
Melting Pt:–7.2 °C
Density:3.12 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-778-1
UN:1744
ADR:8 (6.1),I
 :M-Clarity™ quality level = MQ200
 :GHS06
 :GHS05
 :GHS09
CAS Number:7726-95-6
Formula:Br₂
MDL Number:MFCD00010896