


Bromine ≥99.8%, liquid
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7726-95-6
:
Formula:Br₂
MDL Number:MFCD00010896
:159.81 g/mol
Merck Index:13,01378
Boiling Pt:58.7 °C (1013 hPa)
Melting Pt:–7.2 °C
Density:3.12 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-778-1
UN:1744
ADR:8,I
:GHS06
:GHS05
:GHS09
Related Information
Frequently Bought Together