
Tellurium ≥99.9999% (metals basis), shot, Ø 2-5 mm (0.08-0.20 in)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:13494-80-9
:
Formula:Te
MDL Number:MFCD00134062
:127.6 g/mol
Merck Index:12,09273
Boiling Pt:990 °C (1013 hPa)
Melting Pt:450 °C
Density:8.83 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:236-813-4
UN:3284
ADR:6.1,III
:GHS06
Frequently Bought Together