


Beryllium ≥99.5% (metals basis), foil, Thickness: 0.05 mm (0.002 in)
Catalog # 41613.FF
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:1
- Seq:0001
- Pk Info:25×25 mm
Specifications
About this item
:
:9.01 g/mol
Merck Index:12,01201
Boiling Pt:2970 °C (1013 hPa)
Melting Pt:1278 °C
Density:1.85 g/cm³ (20 °C)
EINECS:231-150-7
:GHS06
:GHS08
:GHS07
Storage Temperature:Ambient
CAS Number:7440-41-7
Formula:Be
MDL Number:MFCD00134032