

Sodium 99.8%, oiled sticks, wrapped in aluminium foil
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7440-23-5
:
Formula:Na
MDL Number:MFCD00085307
:22.99 g/mol
Merck Index:13,08641
Boiling Pt:883 °C (760 mmHg)
Melting Pt:98 °C
Density:0,9 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-132-9
UN:1428
ADR:4.3,I
:GHS02
:GHS05
Specification Test Results
- Appearance:Grey Sticks
- Calcium (Ca):≤400 ppm
- Potassium (K):≤300 ppm
- Sodium (Na):≥99.75 %
Specification Test Results
Frequently Bought Together