

Lithium for synthesis, Sigma-Aldrich®
Catalog # 8.18044.0050
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:50 g
- Seq:3368
Specifications
Specification Test Results
- Solubility:(20 °C) (decomposition)
- Melting Point:179 °C
- Molar Mass:6.94 g/mol
- Boiling Point:1347 °C
- Vapor Pressure:1 hPa (723 °C)
- Density:0.534 g/cm3 (20 °C)
- Hill Formula:Li
- Chemical Formula:Li
Specification Test Results
About this item
:
:6.94 g/mol
Merck Index:13,05542
Boiling Pt:1317 °C (1013 hPa)
Melting Pt:179 °C
Density:0.53 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-102-5
UN:1415
ADR:4.3,I
:M-Clarity™ quality level = MQ200
:GHS02
:GHS05
CAS Number:7439-93-2
Formula:Li
MDL Number:MFCD00134051