
4,4'-Dimethoxybenzophenone ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:90-96-0
:
Formula:C₁₅H₁₄O₃
MDL Number:MFCD00008404
:242.27 g/mol
Boiling Pt:180…240 °C (13 mmHg)
Melting Pt:142…146 °C
Density:1.259 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:202-028-0
:GHS07
Related Information
Frequently Bought Together