Leishman's eosin-methylene blue for histology, stain, Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
Certificates
About this item
CAS Number:12627-53-1
:
MDL Number:MFCD00131498
:M-Clarity™ quality level = MQ100
EINECS:235-732-1
:GHS07
Storage Temperature:Room Temperature
Specification Test Results
- Appearance (Colour):Dark Green to Very Dark Green
- Appearance (Form):Powder
- Infrared spectrum:Conforms to Structure
- Solubility (Colour), c = 1 mg/ml; Methanol:Dark Blue to Very Dark Blue
- Recommended Retest Period:4 Years
Specification Test Results
Frequently Bought Together