Folin-Ciocalteu's reagent for analysis of phenols, Supelco®
: Merck
:
:MFCD00132625
:1.22 g/cm³ (20 °C)
:Ambient
:3264
:8,III
:GHS05
- Equivalent acid:c(H+)= 2 mol/l (2N)
- Sensitivity to phenol:Conforms
- Sensitivity to bovine serum albumin:Conforms
Frequently Bought Together