Order Entry
Italy
ContactUsLinkComponent
X-GlcA ((5-bromo-4-chloro-3-indolyl-ß-D-glucuronide) cyclohexylammonium salt) 98%
X-GlcA ((5-bromo-4-chloro-3-indolyl-ß-D-glucuronide) cyclohexylammonium salt) 98%
Certificates
undefined
X-GlcA ((5-bromo-4-chloro-3-indolyl-ß-D-glucuronide) cyclohexylammonium salt) 98%
Specification Test Results
  • Appearance:
    Colorless to white to off-white Powder or crystals or needles
  • Infrared spectrum:
    Conforms
  • Water:
    ≤5 % (K.F.)
  • HPLC:
    ≥97.5 %