Order Entry
Italy
ContactUsLinkComponent
Thermoreactors, Spectroquant® TR series
Thermoreactors, Spectroquant® TR series
Supplier:  Merck
Certificates
undefined
Thermoreactors, Spectroquant® TR series
Supplier:  Merck
Limited Time Sale
Select items below are currently on sale
Specifications
  • Catalog No:
  • 1.71200.0001
  • 1.71202.0001
  • 1.71201.0001
  • Model:
  • TR320
  • TR620
  • TR420
  • Heating zones:
  • 1
  • 2 separate temperature selectable zones
  • 1
  • Temperature setting:
  • 3 fixed temperatures 100, 120 and 148 ±1 °C
  • Up to 170 ±1 °C
  • Up to 170 ±1 °C
  • Time:
  • As per 8 programs
  • 0 - 180 minutes selectable
  • 0 - 180 minutes, selectable
  • Options:
  • -
  • Thermosensor: Heating block integrated monitoring
  • Thermosensor: Heating block integrated monitoring
  • RS232 connection:
  • -
  • Selectable programs:
  • 8 programs: 148 °C (20 min or 120 min),  150 °C (120 min), 120 °C (30 min, 60 min or 120 min), 100 °C (60 min)
  • 8 pre-set programs as per TR320 plus 8 user-defined programs
  • 8 pre-set programs as per TR320 plus 8 user-defined programs