pH/ORP rod electrodes, SensoLyt®
About this item
The SensoLyt® measuring chains with arming are easy to install and boast flexible measuring and application areas. Eletrodes without arming can be connected directly to a pH monitor of the EcoLine® or QuadroLine® series. Depending on the application, they can be installed into a flow vessel.
- Ideal for measurements in water and drinking water
- Also for high pressure and temperature conditions
- Easy to maintain by easy measuring chain replacement
8 Options Available Below
Product Details & Documents
The SensoLyt® measuring chains with arming are easy to install and boast flexible measuring and application areas. Eletrodes without arming can be connected directly to a pH monitor of the EcoLine® or QuadroLine® series. Depending on the application, they can be installed into a flow vessel.
- Ideal for measurements in water and drinking water
- Also for high pressure and temperature conditions
- Easy to maintain by easy measuring chain replacement
Product Family Options
Product Information
- TypepHForAvailabilityPrice
- 662-0280Sensolyt® SE*Sensolyt® SE*Sensolyt® SE, without arming, for the installation into flow vessels2 - 12Strongly contaminated wastewaterLog in for availability1 * 1 pezzi
- WTWO109102Sensolyt® EC*Sensolyt® EC*Sensolyt® EC, without arming, for the installation into flow vessels2 - 12Normal contaminated waterLog in for availability1 * 1 pezzi
- WTWO109103Sensolyt® DW*Sensolyt® DW*Sensolyt® DW, without arming, to be installed into flow vessels0 - 14Drinking waterLog in for availability1 * 1 pezzi
- 708-0036Sensolyt® SEA pH single rod measuring chain**Sensolyt® SEA pH single rod measuring chain**Sensolyt® SEA, for installation into the SensoLyt® fitting2 - 12Strongly contaminated wastewaterLog in for availability1 * 1 pezzi314,00 €
- 708-0057Sensolyt® ECA pH single rod measuring chain**Sensolyt® ECA pH single rod measuring chain**Sensolyt® ECA, for installation into the SensoLyt® fitting2 - 12Normal contaminated wasterwaterLog in for availability1 * 1 pezzi
- WTWO109118Sensolyt® SEA-HP pH single rod measuring chain**Sensolyt® SEA-HP pH single rod measuring chain**Sensolyt® SEA-HP, to be installed into the SensoLyt® Fitting4 - 12Increased pressure/temperature conditionsLog in for availability1 * 1 pezzi
- 708-0058Sensolyt® DWA pH single rod measuring chain**Sensolyt® DWA pH single rod measuring chain**Sensolyt® DWA, for installation into the SensoLyt® fitting0 - 14Drinking waterLog in for availability1 * 1 pezzi310,00 €
- 708-0037Sensolyt® PtA ORP single rod measuring chain**Sensolyt® PtA ORP single rod measuring chain**Sensolyt® PtA (ORP), for installation into the SensoLyt® fitting±2000 mVStrongly contaminated wastewaterLog in for availability1 * 1 pezzi412,00 €