Order Entry
Italy
Orders LinkContactUsLinkComponent
CryoSleeve™, cryogenic sleeves, Nalgene®
CryoSleeve™, cryogenic sleeves, Nalgene®
Storage tube protection, 273 mm
Catalog # 479-3260
   Low Manufacturing Impact
CryoSleeve™, cryogenic sleeves, Nalgene®
Catalog # 479-3260
   Low Manufacturing Impact
CryoSleeve™, cryogenic sleeves, Nalgene®
CryoSleeve™, cryogenic sleeves, Nalgene®
Storage tube protection, 273 mm
Price
100,00 €
Sale
Valid until 31/03/2026
Log in for availability
1 * 100 pezzi
Product Details & Documents
Sleeve made from PVC encloses aluminium canes with cryotubes for protection during handling and storage.
Specifications
  • Description:
    Storage tube protection, 273 mm
  • Seq:
    16
  • Cat. No.:
    479-3260
  • Environmental certification:
    NAR Certificate of Retirement
  • Environmentally Preferable:
    Low Manufacturing Impact