Screw neck vials, 13-425 thread finish, Chromacol™
Supplier: CHROMACOL
Certificates
About this item
These vials are made from superior quality 1st hydrolytic class clear and amber glass (type 1).
Ordering information: Screw caps need to be ordered separately.
Frequently Bought Together