Order Entry
Italy
Orders LinkContactUsLinkComponent
Sterile swabs
Sterile swabs
Sterile swabs
Supplier:  DELTALAB
Certificates
Sterile swabs
5 Options Available Below
Product Details & Documents
Sterile swabs for samples that do not require a transport container.
Certifications: IVD.
Packaging: Individual peel pouches.
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904
Product Family Options
Product Information
  • Shaft/head
    Ø
    Length
    Sterile
    Availability
    Price
  • Sterile swabs
    Aluminium/cotton
    0,9 mm
    150 mm
    +
    Log in for availability
    Log in for availability
    1 * 1.000 pezzi
    1 * 1.000 pezzi - 830,00 €
    830,00 €
    1 * 1.000 pezzi
       830,00 €
  • Sterile swabs
    Wood/cotton
    2,5 mm
    150 mm
    +
    Log in for availability
    Log in for availability
    1 * 2.000 pezzi
    1 * 2.000 pezzi - 260,00 €
    260,00 €
    1 * 2.000 pezzi
       260,00 €
  • Sterile swabs
    Wood/cotton
    2,5 mm
    150 mm
    +
    Log in for availability
    Log in for availability
    1 * 2.000 pezzi
    1 * 2.000 pezzi - 512,00 €
    512,00 €
    1 * 2.000 pezzi
       512,00 €
  • Sterile swabs
    PS/cotton
    2,5 mm
    150 mm
    +
    Log in for availability
    Log in for availability
    1 * 2.000 pezzi
    1 * 2.000 pezzi - 457,00 €
    457,00 €
    1 * 2.000 pezzi
       457,00 €
  • Sterile swabs
    PS/viscose
    2,2 mm
    200 mm
    +
    Log in for availability
    Log in for availability
    1 * 1.000 pezzi
    1 * 1.000 pezzi - 212,00 €
    212,00 €
    1 * 1.000 pezzi
       212,00 €