Order Entry
Italy
Orders LinkContactUsLinkComponent
Drigalski spatulas
Drigalski spatulas
Supplier:   RSG
Drigalski spatulas
Supplier:  RSG
Certificates
Drigalski spatulas
Drigalski spatulas
3 Options Available Below
Product Details & Documents
Stainless steel 18/10, anti-magnetic.
  • For bacteriological applications
  • Can also be used as stirrer
  • For even spreading or better distribution across media
Product Family Options
Product Information
  • Length
    Blade width
    Availability
    Price
  • 165 mm
    30 mm
    Log in for availability
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 7,00 €
    7,00 €
    1 * 1 pezzi
       7,00 €
  • 165 mm
    40 mm
    Log in for availability
    Log in for availability
    1 * 5 pezzi
    1 * 5 pezzi - 34,80 €
    34,80 €
    1 * 5 pezzi
       34,80 €
  • 195 mm
    40 mm
    Log in for availability
    Log in for availability
    1 * 5 pezzi
    1 * 5 pezzi - 37,60 €
    37,60 €
    1 * 5 pezzi
       37,60 €