Order Entry
Italy
Orders LinkContactUsLinkComponent
VWR®, Refillable Boxes for Next Generation Refills
Refillable racks for 100 - 1250 µl tips, empty (Next Generation)
Catalog # 613-0306
   Low Manufacturing Impact|Sustainable Packaging
Supplier:   VWR Collection
VWR®, Refillable Boxes for Next Generation Refills
Catalog # 613-0306
Supplier:  VWR Collection
   Low Manufacturing Impact|Sustainable Packaging
VWR®, Refillable Boxes for Next Generation Refills
VWR®, Refillable Boxes for Next Generation Refills
Refillable racks for 100 - 1250 µl tips, empty (Next Generation)
Price
Inquire For Price
Log in for availability
1 * 6 pezzi
Product Details & Documents
Hinged refillable autoclavable boxes/racks for pipette tip refills.
  • Virgin resins and sustainable product design
  • Separate insert includes a bevelled top and extra sturdy surface for multi channel pipetting
  • Vented sides ensure effective tip sterilisation during autoclaving
Used with Next Generation refill, Pak Rak and Pak Rak Plus loaders.
Documents
Specifications
  • Description:
    Refillable racks for 100 - 1250 µl tips, empty (Next Generation)
  • Seq:
    202
  • Cat. No.:
    613-0306
  • Environmentally Preferable:
    Low Manufacturing Impact|Sustainable Packaging