Micro haematocrit capillaries, disposable
Supplier: HIRSCHMANN
Certificates
About this item
Disposable capillaries for centrifugation of blood tests.
- Even and heavy wall thickness
- With printed colour-code
According to DIN 12846
Certifications: According to DIN 12846
Packaging: 100 pieces packed in a glass vial, 10 vials in a carton.
Frequently Bought Together