MicroWell™ MiniTrays, low profile, Nunc™
Nome del fornitore: Thermo Fisher Scientific
Certificati
Informazioni su questo articolo
PS, with lid, non sterile. The low profile design and 10 µl well volume make these plates ideal for serological applications.
- Limited adsorption of proteins
- Optical clarity allows use with inverted microscopes
- Lids have keyed tabs for proper orientation
- Large raised alphanumeric coordinates
- Unique surface treatment to maximise sample and reagent mixing
Colour: Clear.
Well dimensions (mm): Top 3,71, Bottom 1,27, Depth 1,91.
External dimensions (mm): 84×59 (with lid).
Specifiche
- Catalog No:
- 732-2751
- 732-2750
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- NAR Certificate of Retirement
- NAR Certificate of Retirement
Specifiche
Frequently Bought Together