Dry box gloves, neoprene
Supplier: PIERCAN
New Product
Certificates
About this item
Ambidextrous gauntlets manufactured from soft black polychloroprene rubber that is self-extinguishing thus providing a good flame resistance for the gauntlet. It is the ideal elastomer providing the best compromise to the conjugate action of several potentially harmful agents, for example oil and ozone. Available for use with various diameter ports.
- High tensile strength and flexibility
- Good resistance to the effects of ozone ageing, oxygen and weathering
Colour: Black
0,4 mm thickness: EN 388 (X100), EN 374-1, EN 421; Cat. III
0,6 mm thickness: EN 388 (X100), EN 374-1 (ACK), EN 421; Cat. III
0,8 mm thickness: EN 388 (X111), EN 374-1 (ACK), EN 421; Cat. III
Certifications: 0,4 mm thickness: EN ISO 374-1 Type B (ANP), EN ISO 374-5 VIRUS, EN 421; Cat. III
0,6 mm thickness: EN 388:2016 (X1XXX), EN ISO 374-1 Type B (ANP), EN ISO 374-5 VIRUS, EN 421; Cat. III
0,8 mm thickness: EN 388:2016 (X111X), EN ISO 374-1 Type B (ANP), EN ISO 374-5 VIRUS, EN 421; Cat. III
0,6 mm thickness: EN 388:2016 (X1XXX), EN ISO 374-1 Type B (ANP), EN ISO 374-5 VIRUS, EN 421; Cat. III
0,8 mm thickness: EN 388:2016 (X111X), EN ISO 374-1 Type B (ANP), EN ISO 374-5 VIRUS, EN 421; Cat. III
Frequently Bought Together