Order Entry
Italy
Orders LinkContactUsLinkComponent
PE gloves
PE gloves
PE gloves
Certificates
PE gloves
2 Options Available Below
Product Details & Documents
Gloves packed in poly bag. Suitable for use in non hazardous environments.
  • Powder-free
  • Textured for a reliable grip
  • Ambidextrous
  • No toxic admixtures, no heavy metal, free from formaldehyde
Colour: Transparent
Specifications for Product Family
Specifications
  • Catalog No:
  • 112-0060
  • 112-0061
  • Ambidextrous / hand-specific:
  • Ambidextrous
  • Ambidextrous
  • Disposable / reusable:
  • Disposable
  • Disposable
  • Powdered / powder-free:
  • Powder-free
  • Powder-free
Specifications
Product Family Options
Product Information
  • Model
    Glove size
    Colour
    Length
    Thickness
    Packed
    Availability
    Price
  • For women
    S
    Transparent
    280 mm
    0,025 mm
    100/bag
    Log in for availability
    Log in for availability
    1 * 100 pezzi
    1 * 100 pezzi - 3,00 €
    3,00 €
    1 * 100 pezzi
       3,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    112-0060
    Ambidextrous / hand-specific
    Ambidextrous
    Disposable / reusable
    Disposable
    Powdered / powder-free
    Powder-free
  • For men
    L
    Transparent
    300 mm
    0,025 mm
    100/bag
    Log in for availability
    Log in for availability
    1 * 100 pezzi
    1 * 100 pezzi - 2,90 €
    2,90 €
    1 * 100 pezzi
       2,90 €
    Specifications:
    More
    Specifications
    Cat. No.
    112-0061
    Ambidextrous / hand-specific
    Ambidextrous
    Disposable / reusable
    Disposable
    Powdered / powder-free
    Powder-free