Nitrile gloves, Solo Ultra 997
: MAPA
These ambidextrous liquidproof gloves with beaded cuffs are made of nitrile.
- Non sterile, chlorinated, textured fingertips
- Excellent dexterity thanks to flexibility and reduced material thickness
- Easy to put on and take off
- Protection of the hand as well as handled products
- Recommended for people sensitised to natural rubber proteins
Ideal for food handling, laboratory work, small parts assembly.
: EN ISO 374-1 Type B (JKT), EN ISO 374-5 VIRUS, Cat. III; AQL 1,5
Food contact approved.
Food contact approved.
- Catalog No:
- 112-0774
- 112-0772
- 112-0771
- 112-0773
- Model:
- 997
- 997
- 997
- 997
- Ambidextrous / hand-specific:
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Disposable / reusable:
- Disposable
- Disposable
- Disposable
- Disposable
- Powdered / powder-free:
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Cuff style:
- Beaded
- Beaded
- Beaded
- Beaded
- EN norm:
- EN ISO 374-1 Type B (JKT), EN ISO 374-5 VIRUS
- EN ISO 374-1 Type B (JKT), EN ISO 374-5 VIRUS
- EN ISO 374-1 Type B (JKT), EN ISO 374-5 VIRUS
- EN ISO 374-1 Type B (JKT), EN ISO 374-5 VIRUS
- Food contact approved:
- +
- +
- +
- +
Frequently Bought Together