Volumetric flasks, class A, blue markings
Supplier: SciLabware
Certificates
About this item
MBL®, borosilicate glass, with smooth neck.
- Inscriptions in blue enamel
- Conformity certified, with batch number
ISO 1042
Certifications: ISO 1042
Frequently Bought Together