Syringe filters, Acrodisc® Versapor® for aqueous samples
Supplier: Cytiva (Formerly Pall Lab)
Certificates
About this item
Useful for prefiltration of particulate laden aqueous samples, serum filtration and dissolution testing.
- Increased pore size for reduced clogging
- Maximized throughput
- Wide range of pore sizes available
Hydrophilic acrylic copolymer membrane; PP housing: 4, 13, Acrodisc® PSF 25 mm; acrylic housing: 25 mm
The glass fiber membrane has very low extractables. Filters are made of 100% binder-free borosilicate glass fiber.
The Versapor® (acrylic copolymer) membrane filters feature large pore sizes ideal for prefiltration. These filters are color-coded by pore size for easy identification.
Polypropylene housing, female Luer-lock inlet and male slip Luer outlet.
Certifications: Acrodisc PSF syringe filters are automation certified to assure smooth operation and worry-free performance 24 hours a day in automated workstations.
Packaging: Syringe filters packed in cases of 200 are available in four packs of 50 or in an AutoPack™ format. The AutoPack™ format consists of eight tubes with 25 syringe filters stacked on top of each other, which is specifically designed for the Caliper™ robotic workstations.
Specifications
- Catalog No:
- 516-1002
- 514-0470
- 516-1001
- 516-1000
- 514-4002
- 514-4104
- 514-0482
- 514-4106
- 514-4024
- 514-4026
- PALL4488
- PALL4473
- Type:
- Premium syringe filter (PSF)
- Premium syringe filter (PSF)
- Premium syringe filter (PSF)
- Syringe filter
- Syringe filter
- Premium syringe filter (PSF)
- Syringe filter
- Syringe filter
- Syringe filter
- Environmentally Preferable:
- Waste Reducing
- Waste Reducing
- Waste Reducing
- Waste Reducing
- Waste Reducing
- Ø:
- 25 mm
- 25 mm
- 25 mm
- 25 mm
- 25 mm
- 25 mm
- 25 mm
- 25 mm
- 25 mm
- 25 mm
- 25 mm
- 4 mm
- Pore size:
- 10,00 µm
- 0,80 µm
- 10,00 µm
- 10,00 µm
- 0,80 µm
- 1,20 µm
- 0,80 µm
- 5,00 µm
- 0,45 µm
- 5,00 µm
- 1,20 µm
- 0,45 µm
- Max. temperature:
- 82 °C
- 82 °C
- 82 °C
- 55 °C
- 55 °C
- 55 °C
- 55 °C
- 55 °C
- 55 °C
- Water flow rate:
- 1182 ml/min (1,0 bar)
- 1182 ml/min (1,0 bar)
- 1182 ml/min (1,0 bar)
- 600 ml/min (3,1 bar)
- 660 ml/min (3,1 bar)
- 905 ml/min (3,1 bar)
- 970 ml/min (3,1 bar)
- 225 ml/min (3,1 bar)
- 970 ml/min (3,1 bar)
Specifications
Related Information
Frequently Bought Together