About this item
Clear PS. Ideal for colony counting.
- Environmentally friendly manufacturing process uses reduced amounts of plastic
- Sterile and particulate-free
90×15 mm dishes
The recessed lid on the nesting and semi-nesting ALPINE dish offers excellent stacking stability. It works with most models of media pouring equipment. The recess on the lid has a smoother bevelled angle allowing the dish to slide easily during automated pouring.
- Vents on the underside of the lid prevent condensation from developing and dripping onto the culture
- Extremely flat bottom allows a reduction in the amount of media required
- Cross ribs on the base make the dish resilient and resistant to warpage when media is poured at a high temperature
- Excellent for performing colony counts, since ribs provide a useful border for making quadrants
85×15 mm space and media saving dishes
Due to its smaller diameter, the 85×15 mm dish is perfect for users who want to save on the amount of media poured, as well as on storage space in the refrigerator and incubator.
90×15 mm sectional dishes
For simultaneous differential studies of organisms or media in a common environment. Overall costs are reduced by minimising use of dishes, media and space. Versions with 3 or 4 sections have numbered compartments for easy identification.
140×15 mm dishes
Ideal for antibiotic susceptibility testing, these dishes accommodate 2 to 3 membrane filters in a single dish. The dish is large enough for bioburden determination and sterility testing of medical devices.
55×15 mm dishes
This dish saves space in refrigerators and incubators, and is ideal when small quantities of media are used. It accommodates single membrane filters for water and pharmaceutical testing.
Deep dishes
Specifically designed to accommodate more media, these dishes are excellent for mycology, histology and pathology labs, as well as for seed germination. Particularly good if longer culturing periods are required.
Speciality dishes
The AQUA PLATE is ideal for membrane filtration techniques. It accommodates a 47 mm round membrane filter, used for a variety of water testing techniques, and is best for field sample collection. The tight fitting lid can be easily sealed with tape. The 40×12,5 mm dish is ideal when only a very small sample is required. It is also suitable for use with small filters and in pharmaceutical analysis. The ALPINE FULL SIZE dish is ideal when a larger surface is required for culture and for accommodating antibiotic sensitivity discs.