Order Entry
Italy
ContactUsLinkComponent
Kidney dishes
Kidney dishes
Catalog # 216-1467
Supplier:  KARTELL
undefined
Kidney dishes
Catalog # 216-1467
Supplier:  KARTELL
Specifications
  • Pk:
    1
Specifications
About this item
PP, white.
  • Very good chemical resistance
  • Very good thermal resistance, autoclavable
  • Smooth surfaces for easy cleaning
These dishes can be used as instrument dishes and for a wide variety of other purposes.
Dimensions: 260×100×55 mm (W×D×H)