Order Entry
Italy
Orders LinkContactUsLinkComponent
Clamps
Clamps
Supplier:   USBECK Laborgeräte
Clamps
Certificates
Clamps
4 Options Available Below
Product Details & Documents
Zinc die-cast, powder coated.
  • The jaws of the clamps are covered with layer of cork to protect the clamped items
  • Without bosshead for fixing to stand
  • With wing screws for rapid easy clamping
Product Family Options
Product Information
  • Spanning width
    Availability
    Price
  • 12 - 25 mm
    Log in for availability
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 11,30 €
    11,30 €
    1 * 1 pezzi
       11,30 €
  • 20 - 40 mm
    Log in for availability
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 12,10 €
    12,10 €
    1 * 1 pezzi
       12,10 €
  • 38 - 60 mm
    Log in for availability
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 13,20 €
    13,20 €
    1 * 1 pezzi
       13,20 €
  • 30 - 80 mm
    Log in for availability
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 19,20 €
    19,20 €
    1 * 1 pezzi
       19,20 €