

1,2,4-Triazole
Catalog # EHERC17649500
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:0,25 g
- Seq:3029
Specifications
About this item
:
:69.07 g/mol
Merck Index:12,09735
Boiling Pt:260 °C (1013 hPa)
Melting Pt:116…119 °C
Density:1.394 g/cm³ (25 °C)
EINECS:206-022-9
:GHS08
:GHS07
CAS Number:288-88-0
Formula:C₂H₃N₃
MDL Number:MFCD00005228