Order Entry
Italy
ContactUsLinkComponent
Methyl blue
Methyl blue
Catalog # ICNA0215264325
Supplier:  MP Biomedicals
CAS Number:  
Methyl blue
Catalog # ICNA0215264325
Supplier:  MP Biomedicals
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
Merck Index:13,06055
 :799.81 g/mol
Storage Temperature:Ambient
EINECS:249-352-9
CAS Number:28983-56-4
Formula:C₃₇H₂₇N₃Na₂O₉S₃
MDL Number:MFCD00058509