Order Entry
Italy
ContactUsLinkComponent
Human Recombinant IL-15 (CTGrade GMP) (from E. coli)
Human Recombinant IL-15 (CTGrade GMP) (from E. coli)
Certificates
undefined
Human Recombinant IL-15 (CTGrade GMP) (from E. coli)

Specifications

  • Conjugation:
    Unconjugated
  • Protein function:
    Cytokine
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Storage conditions:
    At or below –20 °C
  • Endotoxin content:
    Endotoxin LAL (EU/µg) ≤0,05
  • Biological activity:
    Bioactivity Assay: Proliferation of CTLL-2, ED₅₀ ≤1 ng/ml, Bioactivity ≥ 2x10⁴ IU/µg 
  • Gene ID:
    P40933
  • Reconstitution instructions:
    Sterile water at 0,1 - 0,5 mg/ml
  • Endotoxin-free:
    N
  • Carrier-free:
    Y
  • Protease-free:
    N
  • Animal-free:
    Y
  • Protein synonyms:
    IL-T
  • Protein/peptide name:
    CTGrade GMP IL-15
  • Purity:
    ≥97%
  • Molecular weight:
    12,9 kDa
  • Sequence:
    MNWVNVISDL KKIEDLIQSM HIDATLYTES DVHPSCKVTA MKCFLLELQV ISLESGDASI HDTVENLIIL ANNSLSSNGN VTESGCKECE ELEEKNIKEF LQSFVHIVQM FINTS
  • Endotoxin level:
    Low
  • Formulation:
    10 mM Sodium bicarbonate, pH 8,5
  • Nuclease-free:
    N
  • Grade:
    GMP
  • Shipping temperature:
    Ambient
  • Tested applications:
    Bioassay

Specifications

Frequently Bought Together