Order Entry
Italy
ContactUsLinkComponent
Potassium bromate 1/60M (0.01667M) (0.1N) ISO 17034, 17025, 9001
Health
Potassium bromate 1/60M (0.01667M) (0.1N) ISO 17034, 17025, 9001
Catalog # CMPLZ14421053.1L
New Product
CAS Number:  
Health
Potassium bromate 1/60M (0.01667M) (0.1N) ISO 17034, 17025, 9001
Catalog # CMPLZ14421053.1L
New Product
CAS Number:  

Specifications

  • Pk:
    1 L
  • Seq:
    5037

Specifications

About this item

 :
 :167 g/mol
Boiling Pt:100 °C
Melting Pt:0 °C
 :GHS08
Storage Temperature:Room Temperature
CAS Number:7758-01-2
Formula:KBrO₃
MDL Number:MFCD00011359