Sudan IV ≥80% (dye content), certified by the Biological Stain Commission, Sigma-Aldrich®
Catalog # 198102-100G
Supplier: SIGMA ALDRICH MICROSCOPY
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance (Colour):Dark Red to Very Dark Red and Red-Brown
- Appearance (Form):Powder
- Infrared spectrum:Conforms to Structure
- Wavelength:519 - 523 nm
- Extinction coefficient:≥26000
- Biological Stain Commission:Pass
- Dye content:≥80%
Specification Test Results
About this item
Synonyms: Oil Red, 1-((2-Methyl-4-(o-tolyldiazenyl)phenyl)diazenyl)naphthalen-2-ol, Scarlet Red, Oil Red BB, 1-({2-Methyl-4-[(2-methylphenyl)diazenyl]-phenyl}diazenyl)-2-naphthol, Scarlet R, Biebrich scarlet R, Lipid Crimson, Scarlet Red Scharlach, Oil Red IV, Fat Red B, Solvent Red 24, 1-(2-Methyl-4-(2-methylphenylazo)phenylazo)-2-naphthol
:
:380.45 g/mol
EINECS:201-635-8
:GHS07
Storage Temperature:Room Temperature
CAS Number:85-83-6
Formula:C₂₄H₂₀N₄O
MDL Number:MFCD00003893
Melting Pt:199 °C
Merck Index:12,08538
:M-Clarity™ quality level = MQ100