Sudan IV
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Oil Red, 1-((2-Methyl-4-(o-tolyldiazenyl)phenyl)diazenyl)naphthalen-2-ol, Scarlet Red, Oil Red BB, 1-({2-Methyl-4-[(2-methylphenyl)diazenyl]-phenyl}diazenyl)-2-naphthol, Scarlet R, Biebrich scarlet R, Lipid Crimson, Scarlet Red Scharlach, Oil Red IV, Fat Red B, Solvent Red 24, 1-(2-Methyl-4-(2-methylphenylazo)phenylazo)-2-naphthol
CAS Number:85-83-6
:
Formula:C₂₄H₂₀N₄O
MDL Number:MFCD00003893
Molecular Weight:380.45 g/mol
Melting Pt:∼ 185 °C (dec.)
Merck Index:12,08538
EINECS:201-635-8
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together