Order Entry
Italy
ContactUsLinkComponent
1-Octanesulphonic acid sodium salt ≥98.0% for HPLC, for ion pair chromatography, J.T.Baker®
Irritant
1-Octanesulphonic acid sodium salt ≥98.0% for HPLC, for ion pair chromatography, J.T.Baker®
Catalog # 2818-05
Supplier:  Avantor
CAS Number:  
undefined
Irritant
1-Octanesulphonic acid sodium salt ≥98.0% for HPLC, for ion pair chromatography, J.T.Baker®
Catalog # 2818-05
Supplier:  Avantor
CAS Number:  

Specifications

  • Pack type:
    Glass bottle, amber, wide mouth
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • For Laboratory,Research,or Manufacturing Use:
    = 98.0 %
  • Assay (as C₈H₁₇SO₃Na) (by acidimetry):
    ≥98.0 %
  • UV Absorbance of 0.25 M Solution - 250 nm:
    ≤0.050
  • UV Absorbance of 0.25 M Solution - 240 nm:
    ≤0.050
  • UV Absorbance of 0.25 M Solution - 230 nm:
    ≤0.050
  • UV Absorbance of 0.25 M Solution - 220 nm:
    ≤0.060
  • UV Absorbance of 0.25 M Solution - 210 nm:
    ≤0.080
  • UV Absorbance of 0.25 M Solution - 200 nm:
    ≤0.200

Specification Test Results

About this item

 :
 :216.28 g/mol
Melting Pt:> 300 °C
EINECS:226-195-4
 :GHS07
Storage Temperature:Ambient
CAS Number:5324-84-5
Formula:C₈H₁₇NaO₃S
MDL Number:MFCD00007544