Human IFN-gamma Quantikine ELISA Kit
New Product
The Quantikine Human IFN-gamma Immunoassay is a 4.5 hour solid phase ELISA designed to measure human IFN-gamma levels in cell culture supernates, serum, and plasma. It contains HEK293-expressed recombinant human IFN-gamma and antibodies raised against the recombinant factor.
Results obtained for naturally occurring human IFN-gamma samples showed linear curves that were parallel to the standard curves obtained using the Quantikine kit standards. These results indicate that this kit can be used to determine relative mass values for natural human IFN-gamma.
Store the unopened product at 2 to 8 °C.
- Conjugate ELISA:HRP
- Primary antibody reactivity:Human
Frequently Bought Together