4,5-Dichlorophthalic anhydride ≥98.0% (by GC, titration analysis)
Catalog # TCIAD2335-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Purity/Analysis Method:>98.0% (GC,T)
Specification Test Results
About this item
:
:217.01 g/mol
Boiling Pt:313 °C
Melting Pt:188 °C
EINECS:213-386-2
CAS Number:942-06-3
Formula:C₈H₂Cl₂O₃
MDL Number:MFCD00075034