Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Italy
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Benzoin
Benzoin ≥99.0%, zone refined: number of passes 40
Benzoin ≥99.0%, zone refined: number of passes 40
TCIAB0222-1SAMPLE
:
TCI
:
Benzoin ≥99.0%, zone refined: number of passes 40
TCIAB0222-1SAMPLE
:
TCI
:
Pk:
1
Seq:
0014
Purity/Analysis Method:
>99.0% (GC)
:
:
212.25 g/mol
:
344 °C
:
135 °C
:
204-331-3
:
119-53-9
:
C₁₄H₁₂O₂
:
MFCD00004496