Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Italy
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
ε-Caprolactam
ε-Caprolactam ≥99.0%, zone refined: number of passes 24
ε-Caprolactam ≥99.0%, zone refined: number of passes 24
ε-Caprolactam ≥99.0%, zone refined: number of passes 24
TCIAC0022-1SAMPLE
:
TCI
:
ε-Caprolactam ≥99.0%, zone refined: number of passes 24
TCIAC0022-1SAMPLE
:
TCI
:
Pk:
1
Seq:
0014
Purity/Analysis Method:
>99.0% (GC)
:
:
113.16 g/mol
:
267 °C
:
70 °C
:
130 °C
:
203-313-2
:
GHS07
:
105-60-2
:
C₆H₁₁NO
:
MFCD00006936