


Nickel(II) chloride, anhydrous ≥98.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:7718-54-9
:
Formula:Cl₂Ni
MDL Number:MFCD00011142
:129.6 g/mol
Merck Index:12,06588
Melting Pt:1009 °C
Density:3.55 g/cm³ (20 °C)
EINECS:231-743-0
UN:3288
ADR:6.1,III
:GHS06
:GHS08
:GHS09
Storage Temperature:Ambient
Specification Test Results
- Purity/Analysis Method:>98.0% (T)
Specification Test Results
Frequently Bought Together