

Phenanthrene ≥98.5%, zone refined: number of passes 30
Catalog # TCIAP0331-1SAMPLE
Supplier: TCI
CAS Number:
Specifications
- Pk:1
- Seq:0014
Specifications
Specification Test Results
- Purity/Analysis Method:>98.5% (GC)
Specification Test Results
About this item
:
:178.23 g/mol
Merck Index:12,07354
Boiling Pt:340 °C
Melting Pt:100 °C
Density:1.19 g/cm³ (20 °C)
Flash Pt:171 °C
Storage Temperature:Ambient
EINECS:201-581-5
UN:3077
ADR:9,III
:GHS07
:GHS09
CAS Number:85-01-8
Formula:C₁₄H₁₀
MDL Number:MFCD00001168