Imidazole ≥99.8% (by GC), zone refined: number of passes 30
Catalog # TCIAI0014-1SAMPLE
Supplier: TCI
CAS Number:
Specifications
- Pk:1
- Seq:0014
Specifications
Specification Test Results
- Purity/Analysis Method:>99.8% (GC)
Specification Test Results
About this item
:
Melting Pt:92 °C
Flash Pt:145 °C
ADR:8,III
:68.08 g/mol
UN:2923
CAS Number:288-32-4
Formula:C₃H₄N₂
MDL Number:MFCD00005183