


Cobalt(II) chloride hexahydrate ≥98.0% (by titrimetric analysis)
Catalog # TCIAC2388-250G
Supplier: TCI
CAS Number:
Specifications
- Pk:250 g
- Seq:3545
Specifications
Specification Test Results
- Purity/Analysis Method:>98.0% (T)
Specification Test Results
About this item
:
:237.93 g/mol
Merck Index:13,02462
Boiling Pt:110 °C (1013 hPa)
Melting Pt:87 °C
Density:1.92 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-589-4
UN:3077
ADR:9,III
:GHS08
:GHS07
:GHS09
CAS Number:7791-13-1
Formula:CoCl₂·6H₂O
MDL Number:MFCD00149652