Lithium hydroxide, anhydrous ≥98.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:1310-65-2
:
Formula:LiOH
MDL Number:MFCD00011095
:23.95 g/mol
Melting Pt:471 °C
UN:2680
ADR:8,II
Specification Test Results
- Purity/Analysis Method:>98.0% (T)
Specification Test Results
Frequently Bought Together