4-tert-Butylphenol ≥98.0% (by GC), zone refined: number of passes 19
Codice catalogo TCIAB0735-1SAMPLE
Supplier: TCI
CAS Number:
Specifications
- Pk:1
- Seq:0014
Specifications
Specification Test Results
- Purity/Analysis Method:>98.0% (GC)
Specification Test Results
About this item
:
:150.22 g/mol
Boiling Pt:238 °C
Melting Pt:101 °C
Flash Pt:113 °C
UN:2430
ADR:8,III
CAS Number:98-54-4
Formula:C₁₀H₁₄O
MDL Number:MFCD00002367