Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Italy
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Anthracene
Anthracene ≥99.5% (by GC), zone refined: number of passes 30
Anthracene ≥99.5% (by GC), zone refined: number of passes 30
Anthracene ≥99.5% (by GC), zone refined: number of passes 30
TCIAA0405-1SAMPLE
:
TCI
:
Anthracene ≥99.5% (by GC), zone refined: number of passes 30
TCIAA0405-1SAMPLE
:
TCI
:
Pk:
1
Seq:
0014
Purity/Analysis Method:
>99.5% (GC)
:
:
178.23 g/mol
:
342 °C
:
217 °C
:
121 °C
:
120-12-7
:
C₁₄H₁₀
:
MFCD00001240