1,7-Dimethylxanthine 98%
Glass bottle, 250 mg
Catalog # 471832500.
Supplier: Thermo Fisher Scientific
1,7-Dimethylxanthine 98%
Catalog # 471832500.
Supplier: Thermo Fisher Scientific
CAS Number: 611-59-6
Jump to:
Product Details & Documents
Synonyms: Paraxanthine, 1,7-Dimethyl-1H-purine-2,6(3H,7H)-dione
- CAS Number:611-59-6
- Formula:C₇H₈N₄O₂
- MDL Number:MFCD00005727
- Molecular Weight:180.17 g/mol
- Storage Temperature:Ambient
- EINECS:210-271-9
Specifications
Specification Test Results
- Appearance:white to almost white powder
- Infrared spectrum:Conforms
- HPLC:≥97.5 %
- Melting point:294°C to 299°C
Specifications
- Pack type:Glass bottle
- Pk:250 mg



