Order Entry
Italy
ContactUsLinkComponent
Lapatinib ditosylate monohydrate 98%
Irritant
Lapatinib ditosylate monohydrate 98%
Catalog # 469800010.
CAS Number:  
Irritant
Lapatinib ditosylate monohydrate 98%
Catalog # 469800010.
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance:
    yellow crystalline powder
  • Infrared spectrum:
    Conforms
  • HPLC:
    ≥97.5 %
  • Water:
    1.0 to 3.0 %

Specification Test Results

About this item

 :
 :943.48 g/mol
 :GHS07
Storage Temperature:Refrigerator
CAS Number:388082-78-8
Formula:C₄₃H₄₄ClFN₄O₁₁S₃
MDL Number:MFCD18904381