Order Entry
Italy
ContactUsLinkComponent
5,7-Dihydroxy-2-(4-hydroxyphenyl)-6-((2S,3R,4R,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)-4H-chromen-4-one 98%
Irritant
5,7-Dihydroxy-2-(4-hydroxyphenyl)-6-((2S,3R,4R,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)-4H-chromen-4-one 98%
Catalog # 469791000.
CAS Number:  
Irritant
5,7-Dihydroxy-2-(4-hydroxyphenyl)-6-((2S,3R,4R,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)-4H-chromen-4-one 98%
Catalog # 469791000.
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 mg
  • Seq:
    2440

Specifications

Specification Test Results

  • Appearance:
    white to light yellow powder
  • Infrared spectrum:
    Conforms
  • HPLC:
    ≥97.5 %
  • Water:
    ≤5 %

Specification Test Results

About this item

 :
 :432.38 g/mol
 :GHS07
Storage Temperature:Refrigerator
CAS Number:38953-85-4
Formula:C₂₁H₂₀O₁₀
MDL Number:MFCD00076044